Learn More
Biomatik Corporation Xanthine Dehydrogenase (Xdh) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU24058
Encompass_Preferred
Xanthine Dehydrogenase (Xdh) Polyclonal Antibody is a Bovine (Bos taurus) Polyclonal Antibody to Xanthine Dehydrogenase (XDH). The Xanthine Dehydrogenase (Xdh) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Bovine (Bos taurus). Immunogen: Thr2~Leu219. Applications: WB, IHC, ICC, IP. Target Synonyms: Xanthine Dehydrogenase (XDH). Accession Number: P80457. Gene ID: 280960. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9333