Learn More
Biomatik Corporation Iron Responsive Element Binding Protein 2 (Ireb2) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU21869
Encompass_Preferred
Iron Responsive Element Binding Protein 2 (Ireb2) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Iron Responsive Element Binding Protein 2 (IREB2). The Iron Responsive Element Binding Protein 2 (Ireb2) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Pro39~Ser281. Applications: WB, IHC, ICC, IP. Target Synonyms: Iron Responsive Element Binding Protein 2 (IREB2). Accession Number: P48200. Gene ID: 3658. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9226