Learn More
Biomatik Corporation Corin (Crn) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU22294
Encompass_Preferred
Corin (Crn) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Corin (CRN). The Corin (Crn) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Gly70~Thr136. Applications: WB, IHC, ICC, IP. Target Synonyms: Corin (CRN). Accession Number: Q9Y5Q5. Gene ID: 10699. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9250