Learn More
Biomatik Corporation Interleukin 17 Receptor C (Il17Rc) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU23409
Encompass_Preferred
Interleukin 17 Receptor C (Il17Rc) Polyclonal Antibody is a Mouse (Mus musculus) Polyclonal Antibody to Interleukin 17 Receptor C (IL17RC). The Interleukin 17 Receptor C (Il17Rc) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Mouse (Mus musculus). Immunogen: Arg73~Phe330. Applications: WB, IHC, ICC, IP. Target Synonyms: Interleukin 17 Receptor C (IL17RC). Accession Number: Q8K4C2. Gene ID: 171095. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9301