Learn More
Biomatik Corporation 2',5'-Oligoadenylate Synthetase 1 (Oas1) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU23977
Encompass_Preferred
2',5'-Oligoadenylate Synthetase 1 (Oas1) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to 2',5'-Oligoadenylate Synthetase 1 (OAS1). The 2',5'-Oligoadenylate Synthetase 1 (Oas1) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Ser74~Pro165. Applications: WB, IHC, ICC, IP. Target Synonyms: 2',5'-Oligoadenylate Synthetase 1 (OAS1). Accession Number: P00973. Gene ID: 4938. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9328