Learn More
Biomatik Corporation Cytochrome C (Cycs) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU27570
Encompass_Preferred
Cytochrome C (Cycs) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Cytochrome C (CYCS). The Cytochrome C (Cycs) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Glu5~Lys100. Applications: WB, IHC, ICC, IP. Target Synonyms: Cytochrome C (CYCS). Accession Number: P99999. Gene ID: 54205. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9515