Learn More
Biomatik Corporation A Kinase Anchor Protein 11 (Akap11) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU24558
Encompass_Preferred
A Kinase Anchor Protein 11 (Akap11) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to A Kinase Anchor Protein 11 (AKAP11). The A Kinase Anchor Protein 11 (Akap11) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC/IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Leu1650~Ala1901. Applications: WB, IHC, ICC/IF. Target Synonyms: A Kinase Anchor Protein 11 (AKAP11). Accession Number: Q9UKA4. Gene ID: 11215. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9351