Learn More
Biomatik Corporation Kallikrein 8 (Klk8) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU27362
Encompass_Preferred
Kallikrein 8 (Klk8) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Kallikrein 8 (KLK8). The Kallikrein 8 (Klk8) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Gln29~Gly260. Applications: WB, IHC, ICC, IP. Target Synonyms: Kallikrein 8 (KLK8). Accession Number: O60259. Gene ID: 11202. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9504