Learn More
Biomatik Corporation Fatty Acid Binding Protein 4 (Fabp4) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU25486
Encompass_Preferred
Fatty Acid Binding Protein 4 (Fabp4) Polyclonal Antibody is a Chicken (Gallus) Polyclonal Antibody to Fatty Acid Binding Protein 4 (FABP4). The Fatty Acid Binding Protein 4 (Fabp4) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Chicken (Gallus). Immunogen: Met1~Ser125. Applications: WB, IHC, ICC, IP. Target Synonyms: Fatty Acid Binding Protein 4 (FABP4). Accession Number: F1NDE8. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9397