Learn More
Biomatik Corporation Caspase 3 (Casp3) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU27503
Encompass_Preferred
Caspase 3 (Casp3) Polyclonal Antibody is a Rat (Rattus norvegicus) Polyclonal Antibody to Caspase 3 (CASP3). The Caspase 3 (Casp3) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Rat (Rattus norvegicus). Immunogen: Ser29~Asp175. Applications: WB, IHC, ICC, IP. Target Synonyms: Caspase 3 (CASP3). Accession Number: P55213. Gene ID: 25402. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9512