Learn More
Biomatik Corporation Leukocyte Elastase Inhibitor (Lei) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU23375
Encompass_Preferred
Leukocyte Elastase Inhibitor (Lei) Polyclonal Antibody is a Mouse (Mus musculus) Polyclonal Antibody to Leukocyte Elastase Inhibitor (LEI). The Leukocyte Elastase Inhibitor (Lei) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC/IF. Host: Rabbit. Species: Mouse (Mus musculus). Immunogen: Pro184~Ser368. Applications: WB, IHC, ICC/IF. Target Synonyms: Leukocyte Elastase Inhibitor (LEI). Accession Number: Q9D154. Gene ID: 66222. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9299