Learn More
Biomatik Corporation Recombinant Superoxide Dismutase 3, Extracellular (SOD3), 100UG
Supplier:  Biomatik Corporation RPU50349
Encompass_Preferred
Recombinant Human Superoxide Dismutase 3, Extracellular (SOD3) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Superoxide Dismutase 3, Extracellular (SOD3). Target Synonyms: SOD-3; EC-SOD; Extracellular Superoxide Dismutase Cu-Zn. Accession Number: P08294. Expression Region: Trp19~Ala240. Tag Info: N-terminal His Tag. Theoretical MW: 30/33kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5794