Learn More
Biomatik Corporation Recombinant Proopiomelanocortin (POMC), 100UG
Supplier:  Biomatik Corporation RPU51517
Encompass_Preferred
Recombinant Mouse Proopiomelanocortin (POMC) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Proopiomelanocortin (POMC). Target Synonyms: POC; ProOMC; Pro-OMC; Pro Opiomelanocortin; Corticotropin-lipotropin. Accession Number: P01193. Gene ID: 18976. Expression Region: Trp27~Gln235. Tag Info: N-terminal His Tag. Theoretical MW: 33kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5824