Learn More
Biomatik Corporation Recombinant Solute Carrier Family 39, Member 6 (SLC39A6), 100UG
Supplier:  Biomatik Corporation RPU52253
Encompass_Preferred
Recombinant Human Solute Carrier Family 39, Member 6 (SLC39A6) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Solute Carrier Family 39, Member 6 (SLC39A6). Target Synonyms: LIV-1; Zinc Transporter Protein 6; Estrogen-regulated protein LIV-1; Zrt- and Irt-like protein 6; Zinc transporter ZIP6. Accession Number: Q13433. Gene ID: 25800. Expression Region: Phe29~Trp325. Tag Info: N-terminal His Tag. Theoretical MW: 42kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5842