Learn More
Biomatik Corporation Recombinant Interleukin 6 (IL6), 100UG
Supplier:  Biomatik Corporation RPU55173
Encompass_Preferred
Recombinant Human Interleukin 6 (IL6) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Interleukin 6 (IL6). Target Synonyms: MGI2-A; MGI2A; HGF; BSF2; HSF; IFNB2; B-Cell Stimulatory Factor-2; Hybridoma/Plasmacytoma Growth Factor; Hepatocyte Stimulating Factor; Cytotoxic T-Cell Differentiation Factor. Accession Number: P05231. Gene ID: 3569. Expression Region: Pro29~Met212. Tag Info: N-terminal GST Tag. Theoretical MW: 47kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5932