Learn More
Biomatik Corporation Recombinant Phleum pratense Pollen allergen Phl p 5b, 100UG
Supplier:  Biomatik Corporation RPC20305
Encompass_Preferred
Recombinant Phleum pratense Pollen allergen Phl p 5b is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Phleum pratense (Common timothy). Target Name: Phleum pratense Pollen allergen Phl p 5b. Target Synonyms: Allergen Phl p VbAllergen: Phl p 5b. Accession Number: Q40963. Expression Region: 20~284aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 42.1kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4711