Learn More
Biomatik Corporation Recombinant Chlamydia trachomatis Large cysteine-rich periplasmic protein OmcB (omcB), partial, 100UG
Supplier:  Biomatik Corporation RPC20358
Encompass_Preferred
Recombinant Chlamydia trachomatis Large cysteine-rich periplasmic protein OmcB (omcB), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Chlamydia trachomatis (strain D/UW-3/Cx). Target Name: omcB. Target Synonyms: Large-CRP60 kDa cysteine-rich OMP60 kDa outer membrane proteinCysteine-rich outer membrane proteinCRP. Accession Number: P0CC04. Expression Region: 41~196aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 33.1kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4746