Learn More
Biomatik Corporation Recombinant Bacillus subtilis Penicillin-binding protein 4 (pbpD), partial, 100UG
Supplier:  Biomatik Corporation RPC20530
Encompass_Preferred
Recombinant Bacillus subtilis Penicillin-binding protein 4 (pbpD), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Bacillus subtilis (strain 168). Target Name: pbpD. Target Synonyms: pbpD; BSU31490; Penicillin-binding protein 4; PBP 4; Includes: Penicillin-insensitive transglycosylase; EC 2.4.1.129; Peptidoglycan TGase; Penicillin-sensitive transpeptidase; EC 3.4.16.4; DD-transpeptidase. Accession Number: P40750. Expression Region: 213~450aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 43kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4872