Learn More
Biomatik Corporation Recombinant Portulaca grandiflora 4,5-DOPA dioxygenase extradiol (DODA), 100UG
Supplier:  Biomatik Corporation RPC20539
Encompass_Preferred
Recombinant Portulaca grandiflora 4, 5-DOPA dioxygenase extradiol (DODA) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Portulaca grandiflora (Rose moss). Target Name: DODA. Target Synonyms: DODA4,5-DOPA dioxygenase extradiol; EC 1.13.11.29. Accession Number: Q7XA48. Expression Region: 1~271aa. Tag Info: N-Terminal 10Xhis-Sumo-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 49.9kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4880