Learn More
Biomatik Corporation Recombinant Human Malonyl-CoA decarboxylase, mitochondrial (MLYCD), 100UG
Supplier:  Biomatik Corporation RPC20544
Encompass_Preferred
Recombinant Human Malonyl-CoA decarboxylase, mitochondrial (MLYCD) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: MLYCD. Target Synonyms: DCMC_HUMAN; hMCD; Malonyl CoA decarboxylase; Malonyl CoA decarboxylase mitochondrial; Malonyl coenzyme A decarboxylase; Malonyl-CoA decarboxylase; MCD; MGC59795; mitochondrial; Mlycd. Accession Number: O95822. Expression Region: 40~493aa. Tag Info: N-Terminal 10Xhis-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 55.9kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4885