Learn More
Biomatik Corporation Recombinant Mouse Heterogeneous nuclear ribonucleoproteins A2/B1 (Hnrnpa2b1), 100UG
Supplier: Biomatik Corporation RPC20582
Recombinant Mouse Heterogeneous nuclear ribonucleoproteins A2/B1 (Hnrnpa2b1) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Mouse (Mus musculus). Target Name: Hnrnpa2b1. Target Synonyms: hnRNP A2/B1. Accession Number: O88569. Expression Region: 1~353aa. Tag Info: N-Terminal 10Xhis-Sumo-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 57.4kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.