Learn More
Biomatik Corporation Recombinant Mouse Lysosomal acid Glucosylceramidase (Gba), 100UG
Supplier:  Biomatik Corporation RPC21359
Encompass_Preferred
Recombinant Mouse Lysosomal acid Glucosylceramidase (Gba) is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Mouse (Mus musculus). Target Name: Gba. Target Synonyms: Acid beta-glucosidaseBeta-glucocerebrosidaseD-glucosyl-N-acylsphingosine glucohydrolase. Accession Number: P17439. Expression Region: 20~515aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 71.5kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4998