Learn More
Biomatik Corporation Recombinant Vesicular stomatitis Indiana virus RNA-directed RNA polymerase L (L), partial, 100UG
Supplier:  Biomatik Corporation RPC23282
Encompass_Preferred
Recombinant Vesicular stomatitis Indiana virus RNA-directed RNA polymerase L (L), partial is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Vesicular stomatitis Indiana virus (strain 94GUB Central America) (VSIV). Target Name: L. Target Synonyms: Large structural proteinReplicaseTranscriptase. Accession Number: Q8B0H0. Expression Region: 598~784aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 25.4kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5259