Learn More
Biomatik Corporation Recombinant Human Harmonin (USH1C), 100UG
Supplier:  Biomatik Corporation RPC23787
Encompass_Preferred
Recombinant Human Harmonin (USH1C) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: USH1C. Target Synonyms: Antigen NY-CO-38/NY-CO-37Autoimmune enteropathy-related antigen AIE-75Protein PDZ-73Renal carcinoma antigen NY-REN-3Usher syndrome type-1C protein. Accession Number: Q9Y6N9. Expression Region: 1~533aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 76.3kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5297