Learn More
Biomatik Corporation Recombinant Viscum album Viscotoxin-A3 (THI2.1), partial, 100UG
Supplier:  Biomatik Corporation RPC25289
Encompass_Preferred
Recombinant Viscum album Viscotoxin-A3 (THI2.1), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Viscum album (European mistletoe). Target Name: THI2.1. Target Synonyms: THI2.1; Viscotoxin-A3 [Cleaved into: Viscotoxin-A3; Acidic protein]. Accession Number: P01538. Expression Region: 27~72aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 6.8kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5415