Learn More
Biomatik Corporation Recombinant Amyloid Precursor Protein (APP), 100UG
Supplier:  Biomatik Corporation RPU40236
Encompass_Preferred
Recombinant Mouse Amyloid Precursor Protein (APP) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Amyloid Precursor Protein (APP). Target Synonyms: AAA; ABeta; ABPP; AD1; APPI; CVAP; PN2; CTFgamma; Peptidase Nexin-II Alzheimer Disease; Peptidase Nexin-II; Amyloid Beta(A4)Precursor Protein. Accession Number: P12023. Gene ID: 11820. Expression Region: Asp672~Val711. Tag Info: N-terminal His Tag. Theoretical MW: 12kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5461