Learn More
Biomatik Corporation Recombinant Calreticulin (CRT), 100UG
Supplier:  Biomatik Corporation RPU40589
Encompass_Preferred
Recombinant Human Calreticulin (CALR) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Calreticulin (CALR). Target Synonyms: CRT; RO; SSA; CC1qR; ERp60; HACBP; grp60; CRTC; CRP55; Calregulin; Sicca Syndrome Antigen A; Autoantigen Ro; Endoplasmic reticulum resident protein 60. Accession Number: P27797. Gene ID: 811. Expression Region: Glu18~Ser193. Tag Info: N-terminal His Tag. Theoretical MW: 21kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5495