Learn More
Biomatik Corporation Recombinant Defensin Alpha 5, Paneth Cell Specific (DEFa5), 100UG
Supplier:  Biomatik Corporation RPU41153
Encompass_Preferred
Recombinant Human Defensin Alpha 5, Paneth Cell Specific (DEFa5) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Defensin Alpha 5, Paneth Cell Specific (DEFa5). Target Synonyms: DEF-A5; DEF5; HD-5. Accession Number: Q01523. Gene ID: 1670. Expression Region: Glu20~Arg94. Tag Info: N-terminal His Tag. Theoretical MW: 15kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5531