Learn More
Biomatik Corporation Recombinant Human Collagen Type III Alpha 1 (COL3a1)
Supplier:  Biomatik Corporation RPU40895
Encompass_Preferred
Recombinant Human Collagen Type III Alpha 1 (COL3a1) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Collagen Type III Alpha 1 (COL3a1). Target Synonyms: COL3-A1; COL3A1; EDS4A; Collagen Type III Alpha 1; Ehlers-Danlos Syndrome Type IV Autosomal Dominant; Collagen Alpha-1(III)chain. Accession Number: P02461. Gene ID: 1281. Expression Region: Gly1059~Leu1466. Tag Info: N-terminal His Tag. Theoretical MW: 47kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9899