Learn More
Biomatik Corporation Eukaryotic Human Procollagen I N-Terminal Propeptide (PINP)
Supplier:  Biomatik Corporation RPU59029
Encompass_Preferred
Eukaryotic Human Procollagen I N-Terminal Propeptide (PINP) is a purified Eukaryotic Protein. Purity: >95% by SDS-PAGE. Host: 293F Cell. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Procollagen I N-Terminal Propeptide (PINP). Target Synonyms: P1NP; N-Propeptide Of Type I Procollagen; Procollagen I Amino Terminal Propeptide. Accession Number: P02452. Gene ID: 1277. Expression Region: Gln23~Pro161. Tag Info: N-terminal His Tag. Theoretical MW: 32kDa. Buffer: PBS, pH7.4, containing 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0355