Learn More
Biomatik Corporation Recombinant Human Laminin Alpha 1 (LAMA1)
Supplier:  Biomatik Corporation RPU42462
Encompass_Preferred
Recombinant Human Laminin Alpha 1 (LAMA1) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Laminin Alpha 1 (LAMA1). Target Synonyms: LAM-A1; LAMA; Laminin A; LaMa1; S-laminin subunit alpha. Accession Number: P25391. Gene ID: 284217. Expression Region: Ala886~Val1039. Tag Info: N-terminal His Tag. Theoretical MW: 18kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0001