Learn More
Biomatik Corporation Recombinant Human Nucleolin (NCL)
Supplier:  Biomatik Corporation RPU43108
Encompass_Preferred
Recombinant Human Nucleolin (NCL) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Nucleolin (NCL). Target Synonyms: C23; Protein C23. Accession Number: P19338. Gene ID: 4691. Expression Region: Lys388~Gln569. Tag Info: N-terminal His Tag. Theoretical MW: 27kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0029