Learn More
Biomatik Corporation Recombinant Human Glucosidase Alpha, Acid (GaA)
Supplier:  Biomatik Corporation RPU41626
Encompass_Preferred
Recombinant Human Glucosidase Alpha, Acid (GaA) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Glucosidase Alpha, Acid (GaA). Target Synonyms: LYAG; Acid Alpha-Glucosidase; Lysosomal Alpha-Glucosidase; Pompe Disease Glycogen Storage Disease Type II; Acid Maltase; Aglucosidase Alfa. Accession Number: P10253. Gene ID: 2548. Expression Region: Pro595~Gly770. Tag Info: N-terminal His Tag. Theoretical MW: 21kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9956