Learn More
Biomatik Corporation Recombinant Pig Interferon Beta (IFNb)
Supplier:  Biomatik Corporation RPU42133
Encompass_Preferred
Recombinant Pig Interferon Beta (IFNb) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Pig (Sus scrofa; Porcine). Target Name: Interferon Beta (IFNb). Target Synonyms: IFNB1; IFN-B; IFB; IFF; IFNB; Interferon Beta 1 Fibroblast. Accession Number: Q29412. Expression Region: Met22~Asn186. Tag Info: N-terminal His and GST Tag. Theoretical MW: 50kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9987