Learn More
Biomatik Corporation Recombinant Mouse Prion Protein (PRNP)
Supplier:  Biomatik Corporation RPU43413
Encompass_Preferred
Recombinant Mouse Prion Protein (PRNP) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Prion Protein (PRNP). Target Synonyms: CD230; GSS; ASCR; CJD; PRIP; PrP; PrP2730; PrPc; Major prion protein; Creutzfeldt-Jakob Disease; Gerstmann-Strausler-Scheinker Syndrome; Fatal Familial Insomnia. Accession Number: P04925. Gene ID: 19122. Expression Region: Lys23~Ser230. Tag Info: N-terminal His Tag. Theoretical MW: 27kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0043