Learn More
Biomatik Corporation Recombinant Human Acyloxyacyl Hydrolase (AOAH)
Supplier:  Biomatik Corporation RPU52343
Encompass_Preferred
Recombinant Human Acyloxyacyl Hydrolase (AOAH) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Acyloxyacyl Hydrolase (AOAH). Accession Number: P28039. Gene ID: 313. Expression Region: Ser24~Gln300. Tag Info: N-terminal His Tag. Theoretical MW: 35kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0183