Learn More
Biomatik Corporation Recombinant Human Immunoglobulin E (IgE)
Supplier:  Biomatik Corporation RPU55549
Encompass_Preferred
Recombinant Human Immunoglobulin E (IgE) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Immunoglobulin E (IgE). Target Synonyms: IGHE; Immunoglobulin Heavy Constant Epsilon; Ig epsilon chain C region. Accession Number: P01854. Expression Region: Ala1~Lys428. Tag Info: N-terminal His Tag. Theoretical MW: 51kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0307