Learn More
Biomatik Corporation Recombinant Mouse Serum amyloid A-3 protein (Saa3), partial
Supplier:  Biomatik Corporation RPC20133
Encompass_Preferred
Recombinant Mouse Serum amyloid A-3 protein (Saa3), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Mouse (Mus musculus). Target Name: Saa3. Target Synonyms: Saa3; Serum amyloid A-3 protein. Accession Number: P04918. Expression Region: 20~122aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 15.8kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0378