Learn More
Biomatik Corporation Recombinant Human Cancer/testis antigen 1 (CTAG1A)
Supplier:  Biomatik Corporation RPC20225
Encompass_Preferred
Recombinant Human Cancer/testis antigen 1 (CTAG1A) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: CTAG1A. Target Synonyms: Autoimmunogenic cancer/testis antigen NY-ESO-1Cancer/testis antigen 6.1; CT6.1L antigen family member 2; LAGE-2. Accession Number: P78358. Expression Region: 1~180aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 20kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0387