Learn More
Biomatik Corporation Recombinant Escherichia coli UDP-3-O-acyl-N-acetylglucosamine deacetylase (lpxC)
Supplier:  Biomatik Corporation RPC20461
Encompass_Preferred
Recombinant Escherichia coli UDP-3-O-acyl-N-acetylglucosamine deacetylase (lpxC) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Escherichia coli (strain K12). Target Name: lpxC. Target Synonyms: Protein EnvAUDP-3-O-[R-3-hydroxymyristoyl]-N-acetylglucosamine deacetylase. Accession Number: P0A725. Expression Region: 1~305aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 38kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0410