Learn More
Biomatik Corporation Recombinant Human Deoxyribonuclease II (DNASEII)
Supplier:  Biomatik Corporation RPU51789
Encompass_Preferred
Recombinant Human Deoxyribonuclease II (DNASEII) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Deoxyribonuclease II (DNASEII). Target Synonyms: DNase-II; DNase2; DNASE2A; DNASE2B; DRN2; DNL; DNL2; Deoxyribonuclease II Alpha; Acid DNase; Lysosomal DNase II. Accession Number: O00115. Gene ID: 1777. Expression Region: Cys19~Ile360. Tag Info: N-terminal His Tag. Theoretical MW: 42kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0160