Learn More
Biomatik Corporation Native Bovine Beta-Lactoglobulin (bLg)
Supplier:  Biomatik Corporation RPU55153
Encompass_Preferred
Native Bovine Beta-Lactoglobulin (bLg) is a purified Natural Protein. Purity: >90% by SDS-PAGE. Host: Bovine. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Bovine (Bos taurus). Target Name: Beta-Lactoglobulin (bLg). Target Synonyms: LGB; B-LG. Accession Number: P02754. Gene ID: 280838. Theoretical MW: 18kDa. Buffer: PBS, pH7.4, containing 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0278