Learn More
Biomatik Corporation Recombinant Bovine Casein Alpha (CSN1)
Supplier:  Biomatik Corporation RPU53691
Encompass_Preferred
Recombinant Bovine Casein Alpha (CSN1) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Bovine (Bos taurus). Target Name: Casein Alpha (CSN1). Target Synonyms: CASa; CSN1; CSN1S1; Casein Alpha S1; Caseinate; Caseine/caséine/Kasein/caseína; Alpha-S1-casein; Casoxin-D. Accession Number: P02662. Gene ID: 282208. Expression Region: Arg16~Gln187. Tag Info: N-terminal His Tag. Theoretical MW: 30kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0231