Learn More
Biomatik Corporation Recombinant Human Cereblon (CRBN)
Supplier:  Biomatik Corporation RPU56160
Encompass_Preferred
Recombinant Human Cereblon (CRBN) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Cereblon (CRBN). Target Synonyms: MRT2A; Mental Retardation,Non-Syndromic,Autosomal Recessive 2A. Accession Number: Q96SW2. Gene ID: 51185. Expression Region: Lys300~Glu430. Tag Info: N-terminal His Tag. Theoretical MW: 20kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0328