Learn More
Biomatik Corporation Recombinant Human Eosinophil Peroxidase (EPX)
Supplier:  Biomatik Corporation RPU58582
Encompass_Preferred
Recombinant Human Eosinophil Peroxidase (EPX) is a purified Prokaryotic Protein. Purity: >80% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Eosinophil Peroxidase (EPX). Target Synonyms: EPO; EPP; EPX-PEN; EPER. Accession Number: P11678. Gene ID: 8288. Expression Region: Arg140~Thr715. Tag Info: N-terminal His Tag. Theoretical MW: 70/28kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0352