Learn More
Biomatik Corporation Recombinant Human Amelogenin, X isoform (AMELX)
Supplier:  Biomatik Corporation RPC20374
Encompass_Preferred
Recombinant Human Amelogenin, X isoform (AMELX) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: AMELX. Target Synonyms: AI1E; AIH1; ALGN; Amel; Amelogenesis imperfecta 1; Amelogenin (amelogenesis imperfecta 1; X linked); Amelogenin (X chromosome); Amelogenin (X chromosome; amelogenesis imperfecta 1); Amelogenin; Amelogenin X isoform; Amelogenin; X linked; AMELX; AMELX_HUMAN; Amg. Accession Number: Q99217. Expression Region: 17~191aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 35.9kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0406