Learn More
Biomatik Corporation Recombinant Human Cytokeratin 10 (CK10)
Supplier:  Biomatik Corporation RPU42376
Encompass_Preferred
Recombinant Human Cytokeratin 10 (CK10) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Cytokeratin 10 (CK10). Target Synonyms: K10; KRT10; KPP; Keratin 10; Epidermolytic Hyperkeratosis; Keratosis Palmaris Et Plantaris; Keratin, type I cytoskeletal 10. Accession Number: P13645. Gene ID: 3858. Expression Region: Leu153~Glu456. Tag Info: N-terminal His Tag. Theoretical MW: 40kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9998