Learn More
Biomatik Corporation Recombinant Human Mucin 6 (MUC6)
Supplier:  Biomatik Corporation RPU42838
Encompass_Preferred
Recombinant Human Mucin 6 (MUC6) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Mucin 6 (MUC6). Target Synonyms: Mucin 6,Gastric; Oligomeric Mucus/Gel-Forming. Accession Number: Q6W4X9. Expression Region: Leu2338~Asp2439. Tag Info: N-terminal His and GST Tag. Theoretical MW: 41kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0019