Learn More
Biomatik Corporation Recombinant Human Protein Disulfide Isomerase (PDI)
Supplier:  Biomatik Corporation RPU43476
Encompass_Preferred
Recombinant Human Protein Disulfide Isomerase (PDI) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Protein Disulfide Isomerase (PDI). Target Synonyms: P4HB; ERBA2L; PDIA1; PO4DB; PROHB, DSI, GIT, PDI, PO4HB, P4Hbeta; Prolyl 4-Hydroxylase Subunit Beta; Procollagen-Proline 2-Oxoglutarate 4-Dioxygenase Beta Polypeptide. Accession Number: P07237. Gene ID: 5034. Expression Region: Phe216~Gly465. Tag Info: N-terminal His and GST Tag. Theoretical MW: 58/55kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0048