Learn More
Sino Biological Peptide Substrates: CATCHtide
Supplier:  Sino Biological C0258
Encompass_Preferred
The CATCHtide peptide sequence (CRRHYYYDTHTNTYY-LRTFGHNTRR) is derived from human SLC12A2 (amino acids 198-217) and is suitable as a substrate for kinases OSR1 and STK39.
Catalog No. 50-256-9656